Product Details

SNP ID
rs112696197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103277688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCAGCTGGAGGAGGCCAGCTC[G/T]GCTCGCTCGCAGGCCCTGCTGGAGC
Phenotype
MIM: 605338
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
INA PubMed Links

Gene Details

Gene
INA
Gene Name
internexin neuronal intermediate filament protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032727.3 526 Silent Mutation TCG,TCT S159S NP_116116.1

View Full Product Details