Product Details

SNP ID
rs143184593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15105643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTCATGGATTATGCCATCTCT[C/T]GATTTAAATTTTACTTAGTGCCATC
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf111 PubMed Links

Gene Details

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
There are no transcripts associated with this gene.

Gene
NMT2
Gene Name
N-myristoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308295.1 Intron NP_001295224.1
NM_004808.2 Intron NP_004799.1
XM_005252642.4 Intron XP_005252699.1
XM_006717539.3 Intron XP_006717602.1
XM_017016947.1 Intron XP_016872436.1
XM_017016948.1 Intron XP_016872437.1
XM_017016949.1 Intron XP_016872438.1
XM_017016950.1 Intron XP_016872439.1
XM_017016951.1 Intron XP_016872440.1
XM_017016952.1 Intron XP_016872441.1
XM_017016953.1 Intron XP_016872442.1
Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
There are no transcripts associated with this gene.

View Full Product Details