Product Details
- SNP ID
-
rs148502124
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:114188004 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCTAATTTCAGGCTGAAAAGCT[C/T]AGAGAATGGAAATAAAAAGAACAAT
- Phenotype
-
MIM: 605796
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TDRD1
PubMed Links
Gene Details
- Gene
- TDRD1
- Gene Name
- tudor domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198795.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
NP_942090.1 |
XM_005269978.2 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_005270035.1 |
XM_005269979.2 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_005270036.1 |
XM_011539959.2 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538261.1 |
XM_011539960.2 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538262.1 |
XM_011539961.2 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538263.1 |
XM_011539962.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538264.1 |
XM_011539963.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538265.1 |
XM_011539964.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538266.1 |
XM_011539966.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_011538268.1 |
XM_017016414.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_016871903.1 |
XM_017016415.1 |
340 |
Missense Mutation |
TCA,TTA |
S58L |
XP_016871904.1 |
View Full Product Details