Product Details

SNP ID
rs150065176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:129841683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCACTGTGGGATGGGGTGCGG[A/G]GTGGGGAAATGGGGGTCTGTCTCCA
Phenotype
MIM: 607407
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EBF3 PubMed Links

Gene Details

Gene
EBF3
Gene Name
early B-cell factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005463.2 Intron NP_001005463.1
XM_005252667.3 Intron XP_005252724.1
XM_005252668.3 Intron XP_005252725.1
XM_005252669.3 Intron XP_005252726.1
XM_006717739.3 Intron XP_006717802.1
XM_006717740.3 Intron XP_006717803.1
XM_006717741.3 Intron XP_006717804.1
XM_006717742.3 Intron XP_006717805.1
XM_006717743.3 Intron XP_006717806.1
XM_006717744.3 Intron XP_006717807.1
XM_011539574.2 Intron XP_011537876.1
XM_011539575.2 Intron XP_011537877.1
XM_017016027.1 Intron XP_016871516.1
Gene
MIR4297
Gene Name
microRNA 4297
There are no transcripts associated with this gene.

View Full Product Details