Product Details

SNP ID
rs150116466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27110716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTTTGAAACCAAGCACAGTTA[G/T]AGTCAAAATGAGGGACATTATTCAG
Phenotype
MIM: 607472
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YME1L1 PubMed Links

Gene Details

Gene
YME1L1
Gene Name
YME1 like 1 ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253866.1 4076 UTR 3 NP_001240795.1
NM_014263.3 4076 UTR 3 NP_055078.1
NM_139312.2 4076 UTR 3 NP_647473.1
XM_011519300.2 4076 UTR 3 XP_011517602.1

View Full Product Details