Product Details

SNP ID
rs117013478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78216520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCTTTCAAAAGGTCAATTTCA[C/G]CTCTCATCCTTCCATCTCTACCCCT
Phenotype
MIM: 606203
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GAB2 PubMed Links

Gene Details

Gene
GAB2
Gene Name
GRB2 associated binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012296.3 4885 UTR 3 NP_036428.1
NM_080491.2 4885 UTR 3 NP_536739.1
XM_006718753.2 4885 UTR 3 XP_006718816.1
XM_011545408.2 4885 UTR 3 XP_011543710.1
Gene
USP35
Gene Name
ubiquitin specific peptidase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020798.2 4885 Intron NP_065849.1
XM_011545187.2 4885 Intron XP_011543489.1
XM_011545188.2 4885 Intron XP_011543490.1
XM_017018052.1 4885 Intron XP_016873541.1
XM_017018053.1 4885 Intron XP_016873542.1

View Full Product Details