Product Details

SNP ID
rs137923849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6877008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGCCTCCTGGCCACCATGGCA[A/T]ATGACCGCTACGTGGCCATCTGTGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10A2 PubMed Links

Gene Details

Gene
OR10A2
Gene Name
olfactory receptor family 10 subfamily A member 2
There are no transcripts associated with this gene.

Gene
OR10A4
Gene Name
olfactory receptor family 10 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207186.2 384 Missense Mutation AAT,TAT N121Y NP_997069.2

View Full Product Details