Product Details

SNP ID
rs138493180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18532458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGGATGCAAGGCAAACTTAA[A/G]AACACTTCACTATTTATATCATAAT
Phenotype
MIM: 601387 MIM: 610985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSG101 PubMed Links

Gene Details

Gene
TSG101
Gene Name
tumor susceptibility 101
There are no transcripts associated with this gene.

Gene
UEVLD
Gene Name
UEV and lactate/malate dehyrogenase domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040697.2 711 Silent Mutation TTC,TTT F426F NP_001035787.1
NM_001261382.1 711 Silent Mutation TTC,TTT F404F NP_001248311.1
NM_001261383.1 711 UTR 3 NP_001248312.1
NM_001261384.1 711 Silent Mutation TTC,TTT F296F NP_001248313.1
NM_001261385.1 711 UTR 3 NP_001248314.1
NM_001261386.1 711 UTR 3 NP_001248315.1
NM_001297771.1 711 Intron NP_001284700.1
NM_018314.4 711 UTR 3 NP_060784.3
XM_006718255.2 711 Silent Mutation TTC,TTT F203F XP_006718318.1
XM_011520201.2 711 Silent Mutation TTC,TTT F203F XP_011518503.1
XM_017017985.1 711 Silent Mutation TTC,TTT F296F XP_016873474.1
XM_017017986.1 711 UTR 3 XP_016873475.1

View Full Product Details