Product Details

SNP ID
rs138794116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65655754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGTCGAGGTGGAGGCCGGGGGT[C/T]GGTGGGTCCTGTAGGGCAAGGGCTA
Phenotype
MIM: 164014 MIM: 602180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4489 PubMed Links

Gene Details

Gene
MIR4489
Gene Name
microRNA 4489
There are no transcripts associated with this gene.

Gene
RELA
Gene Name
RELA proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145138.1 875 Missense Mutation AAC,GAC N320D NP_001138610.1
NM_001243984.1 875 Missense Mutation AAC,GAC N323D NP_001230913.1
NM_001243985.1 875 Missense Mutation AAC,GAC N323D NP_001230914.1
NM_021975.3 875 Missense Mutation AAC,GAC N323D NP_068810.3
XM_011545206.1 875 Missense Mutation AAC,GAC N252D XP_011543508.1
XM_011545207.1 875 Missense Mutation AAC,GAC N217D XP_011543509.1
Gene
SIPA1
Gene Name
signal-induced proliferation-associated 1
There are no transcripts associated with this gene.

View Full Product Details