Product Details

SNP ID
rs140206085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116790381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGCGGAAAGCCTGAAGTCGC[C/T]GGCGCACCTCCTCGGAGAGCATCTG
Phenotype
MIM: 606368 MIM: 603901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOA5 PubMed Links

Gene Details

Gene
APOA5
Gene Name
apolipoprotein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166598.1 921 Missense Mutation CAG,CGG Q283R NP_001160070.1
NM_052968.4 921 Missense Mutation CAG,CGG Q283R NP_443200.2
XM_017017169.1 921 Missense Mutation CAG,CGG Q283R XP_016872658.1
Gene
ZPR1
Gene Name
ZPR1 zinc finger
There are no transcripts associated with this gene.

View Full Product Details