Product Details

SNP ID
rs141217844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64556074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCTGCAGGTGCTCACCTTCAT[C/T]CTCCCCTGCCTCATGATACCTTCCC
Phenotype
MIM: 607097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A11 PubMed Links

Gene Details

Gene
SLC22A11
Gene Name
solute carrier family 22 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307985.1 474 Silent Mutation ATC,ATT I25I NP_001294914.1
NM_018484.3 474 Silent Mutation ATC,ATT I25I NP_060954.1
XM_011545167.1 474 UTR 5 XP_011543469.1

View Full Product Details