Product Details

SNP ID
rs142967171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72577393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCCTGGAGGGGATCACTCAGC[A/T]TCAAGGCTGCAGCAGCCATTGATGG
Phenotype
MIM: 602658
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 3296 Missense Mutation GAA,GAT E932D NP_001137311.1
NM_001146209.2 3296 Missense Mutation GAA,GAT E930D NP_001139681.1
NM_001243784.1 3296 Missense Mutation GAA,GAT E918D NP_001230713.1
NM_002599.4 3296 Missense Mutation GAA,GAT E939D NP_002590.1
XM_005274040.2 3296 Missense Mutation GAA,GAT E938D XP_005274097.1
XM_011545096.2 3296 Missense Mutation GAA,GAT E618D XP_011543398.1
XM_017017910.1 3296 Missense Mutation GAA,GAT E618D XP_016873399.1

View Full Product Details