Product Details
- SNP ID
-
rs143114305
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:65946997 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGGCCAGAGCAAGAAAGGACAG[G/T]GCTCTGAAGAGTCTGAAGAGTAAGC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TSGA10IP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7926657] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TSGA10IP
- Gene Name
- testis specific 10 interacting protein
View Full Product Details