Product Details
- SNP ID
-
rs143648713
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102772867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACCTCCAGTATTTGTCCGCTGC[A/C]AAGAAGTATGTTTTCTTCTTTTCCT
- Phenotype
-
MIM: 185260
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MMP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17860992] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MMP10
- Gene Name
- matrix metallopeptidase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002425.2 |
1243 |
Missense Mutation |
TTG,TTT |
L402F |
NP_002416.1 |
View Full Product Details