Product Details

SNP ID
rs147616072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60919869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGATGTCTTGACCCAGATAGGTC[A/G]ATCTGTGCGAGGGACACTGGATGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM109 PubMed Links

Gene Details

Gene
TMEM109
Gene Name
transmembrane protein 109
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024092.2 554 Missense Mutation CAA,CGA Q59R NP_076997.1
Gene
TMEM132A
Gene Name
transmembrane protein 132A
There are no transcripts associated with this gene.

View Full Product Details