Product Details

SNP ID
rs148133757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72578991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGAAGTCTGTGGTGCTGCTTG[C/T]TGTTTCGGTCGTAGCCCACTGTTGA
Phenotype
MIM: 602658
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 2854 Missense Mutation AAC,AGC N785S NP_001137311.1
NM_001146209.2 2854 Missense Mutation AAC,AGC N783S NP_001139681.1
NM_001243784.1 2854 Missense Mutation AAC,AGC N771S NP_001230713.1
NM_002599.4 2854 Missense Mutation AAC,AGC N792S NP_002590.1
XM_005274040.2 2854 Missense Mutation AAC,AGC N791S XP_005274097.1
XM_011545096.2 2854 Missense Mutation AAC,AGC N471S XP_011543398.1
XM_017017910.1 2854 Missense Mutation AAC,AGC N471S XP_016873399.1

View Full Product Details