Product Details

SNP ID
rs148855440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4803905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACCATGAAGCAGAAGGGGCTC[A/G]CCCACAGCAGCCCTCTCAGCATCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52R1 PubMed Links

Gene Details

Gene
OR52R1
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005177.3 476 Missense Mutation GCG,GTG A159V NP_001005177.3

View Full Product Details