Product Details

SNP ID
rs141193184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108787100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATCTGGGCTCTCAGCAGGACGA[C/T]GTGTCTCAGAACCACCACCTGAGCC
Phenotype
MIM: 606778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSH1 PubMed Links

Gene Details

Gene
SSH1
Gene Name
slingshot protein phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161330.1 3922 Intron NP_001154802.1
NM_001161331.1 3922 Intron NP_001154803.1
NM_018984.3 3922 UTR 3 NP_061857.3
XM_005268984.4 3922 UTR 3 XP_005269041.1
XM_005268985.2 3922 UTR 3 XP_005269042.1
XM_011538497.1 3922 UTR 3 XP_011536799.1
XM_011538499.1 3922 UTR 3 XP_011536801.1
XM_011538500.2 3922 UTR 3 XP_011536802.1
XM_011538501.2 3922 UTR 3 XP_011536803.1
XM_017019491.1 3922 UTR 3 XP_016874980.1
XM_017019492.1 3922 UTR 3 XP_016874981.1
XM_017019493.1 3922 UTR 3 XP_016874982.1
XM_017019494.1 3922 UTR 3 XP_016874983.1

View Full Product Details