Product Details

SNP ID
rs142808589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48830542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTATGCTGGGCATCTGGGTGG[C/T]TGGCTAGTTCGGGGGGACAGGAAGA
Phenotype
MIM: 601958 MIM: 612172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNB3 PubMed Links

Gene Details

Gene
CACNB3
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
There are no transcripts associated with this gene.

Gene
DDX23
Gene Name
DEAD-box helicase 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004818.2 Intron NP_004809.2

View Full Product Details