Product Details

SNP ID
rs145170340
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50058802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGAGGAGGAGGAGGTGGGTGG[C/T]GTCCAGCCGGTGAGCATCCAGGCCT
Phenotype
MIM: 602866
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ASIC1 PubMed Links

Gene Details

Gene
ASIC1
Gene Name
acid sensing ion channel subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001095.3 115 Silent Mutation GGC,GGT G12G NP_001086.2
NM_001256830.1 115 Intron NP_001243759.1
NM_020039.3 115 Silent Mutation GGC,GGT G12G NP_064423.2
XM_011538350.1 115 Silent Mutation GGC,GGT G12G XP_011536652.1
XM_011538351.2 115 Silent Mutation GGC,GGT G12G XP_011536653.1
XM_011538352.1 115 Silent Mutation GGC,GGT G12G XP_011536654.1

View Full Product Details