Product Details
- SNP ID
-
rs146403821
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:107775717 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCGGCGCCTTCGCCCAGCAGCGAG[A/C]CCGAGGAGGGGGGCAGCTAGCGAGC
- Phenotype
-
MIM: 609155
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ASCL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201513808] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASCL4
- Gene Name
- achaete-scute family bHLH transcription factor 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203436.2 |
1333 |
Missense Mutation |
ACC,CCC |
T168P |
NP_982260.2 |
View Full Product Details