Product Details

SNP ID
rs147582132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123584774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGGGCACCAAGATGGGCCTCAT[C/T]TTCGAGGTGGCGGAGGGCGGCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927415 PubMed Links

Gene Details

Gene
LOC101927415
Gene Name
uncharacterized LOC101927415
There are no transcripts associated with this gene.

Gene
TMED2
Gene Name
transmembrane p24 trafficking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321445.1 246 Silent Mutation ATC,ATT I46I NP_001308374.1
NM_006815.3 246 Silent Mutation ATC,ATT I46I NP_006806.1

View Full Product Details