Product Details

SNP ID
rs147856424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52692499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGCTTGACCCCCAGCATGGCC[C/T]GGTAGTCACGCAGCAGCCGGGCCAG
Phenotype
MIM: 611158
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT77 PubMed Links

Gene Details

Gene
KRT77
Gene Name
keratin 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175078.2 1140 Missense Mutation CAG,CGG Q450R NP_778253.2
XM_011538288.2 1140 Missense Mutation CAG,CGG Q217R XP_011536590.1
XM_011538289.2 1140 Intron XP_011536591.1

View Full Product Details