Product Details

SNP ID
rs148154047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48981534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGCGGCAGGGGCCACCGCA[C/T]GCGCACGCAGCGCGTCACCGAGCGC
Phenotype
MIM: 164820 MIM: 601906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT1 PubMed Links

Gene Details

Gene
WNT1
Gene Name
Wnt family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005430.3 1205 Missense Mutation ACG,ATG T336M NP_005421.1
Gene
WNT10B
Gene Name
Wnt family member 10B
There are no transcripts associated with this gene.

View Full Product Details