Product Details

SNP ID
rs149668192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120694218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCATCAAAGAGGAGGGGGACTA[C/T]GTGCTGGTCTTGAAATTTGCAGAGG
Phenotype
MIM: 613802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MLEC PubMed Links

Gene Details

Gene
MLEC
Gene Name
malectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303627.1 536 Silent Mutation TAC,TAT Y38Y NP_001290556.1
NM_001303628.1 536 Silent Mutation TAC,TAT Y121Y NP_001290557.1
NM_014730.3 536 Silent Mutation TAC,TAT Y121Y NP_055545.1
XM_011539031.1 536 Silent Mutation TAC,TAT Y38Y XP_011537333.1
XM_011539032.1 536 Silent Mutation TAC,TAT Y38Y XP_011537334.1

View Full Product Details