Product Details
- SNP ID
-
rs150334581
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52690074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTTCCAAGACCCAAGTCAGATC[C/T]TCAGACTCCTCAGGCCCGAGGCACC
- Phenotype
-
MIM: 139350
MIM: 611158
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT1
PubMed Links
Gene Details
- Gene
- KRT1
- Gene Name
- keratin 1
There are no transcripts associated with this gene.
- Gene
- KRT77
- Gene Name
- keratin 77
View Full Product Details