Product Details

SNP ID
rs140685040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72759801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTTTGTTGAATATAGCTATTT[C/T]CCAAGCTTCATCTTCTTTTTCTTTG
Phenotype
MIM: 610510 MIM: 607533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
There are no transcripts associated with this gene.

Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128226.2 3167 Silent Mutation GGA,GGG G927G NP_001121698.1
NM_001322348.1 3167 Silent Mutation GGA,GGG G834G NP_001309277.1
NM_001322349.1 3167 Silent Mutation GGA,GGG G795G NP_001309278.1
NM_014953.4 3167 Silent Mutation GGA,GGG G957G NP_055768.3

View Full Product Details