Product Details

SNP ID
rs141067458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72759797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGATTTTTGTTGAATATAGCT[A/G]TTTTCCAAGCTTCATCTTCTTTTTC
Phenotype
MIM: 610510 MIM: 607533
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
There are no transcripts associated with this gene.

Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
There are no transcripts associated with this gene.

View Full Product Details