Product Details

SNP ID
rs148246113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39655845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAACCCGCTGTCGCGCAAGCTGC[A/G]TAAGATCCTGGAGACGCGGCTGGAC
Phenotype
MIM: 606977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG6 PubMed Links

Gene Details

Gene
COG6
Gene Name
component of oligomeric golgi complex 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145079.1 219 Missense Mutation CAT,CGT H40R NP_001138551.1
NM_020751.2 219 Missense Mutation CAT,CGT H40R NP_065802.1
XM_011535168.1 219 Missense Mutation CAT,CGT H40R XP_011533470.1
Gene
MIR4305
Gene Name
microRNA 4305
There are no transcripts associated with this gene.

View Full Product Details