Product Details

SNP ID
rs143160976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73071710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACTCCCACCAGGGATCCCACC[C/T]CCGCAGTTTCCAGGATTTCCTCCAC
Phenotype
MIM: 612427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM25 PubMed Links

Gene Details

Gene
RBM25
Gene Name
RNA binding motif protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021239.2 277 Silent Mutation CCC,CCT P23P NP_067062.1
XM_011537044.2 277 Silent Mutation CCC,CCT P23P XP_011535346.1
XM_011537045.2 277 UTR 5 XP_011535347.1

View Full Product Details