Product Details

SNP ID
rs144815201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22873366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGCAGGGCACCTTACAGAGGCC[C/T]CTGGTCCGGGACAGCCGCACCTCCC
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267510.1 826 Silent Mutation CCC,CCT P45P XP_005267567.1
Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
There are no transcripts associated with this gene.

View Full Product Details