Product Details

SNP ID
rs144981852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94563767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCTTTCCCTGGGGGCCTGCTC[A/T]CACAGCCGCAGCCAGATCCTTGAGG
Phenotype
MIM: 147935
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA4 PubMed Links

Gene Details

Gene
SERPINA4
Gene Name
serpin family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289032.1 377 Silent Mutation TCA,TCT S132S NP_001275961.1
NM_001289033.1 377 Silent Mutation TCA,TCT S95S NP_001275962.1
NM_006215.3 377 Silent Mutation TCA,TCT S95S NP_006206.2

View Full Product Details