Product Details

SNP ID
rs145103222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94563576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGAGCATGATGGTGAGAGTTGC[A/G]GTAACAGCTCCCACCAGCAGATTCT
Phenotype
MIM: 147935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA4 PubMed Links

Gene Details

Gene
SERPINA4
Gene Name
serpin family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289032.1 186 Missense Mutation AGT,GGT S69G NP_001275961.1
NM_001289033.1 186 Missense Mutation AGT,GGT S32G NP_001275962.1
NM_006215.3 186 Missense Mutation AGT,GGT S32G NP_006206.2

View Full Product Details