Product Details
- SNP ID
-
rs149443791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:60712939 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTGTGCATATATATATGCATG[C/T]TACTTAACTTTAAAAATATGTACCA
- Phenotype
-
MIM: 606342
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SIX4
PubMed Links
Gene Details
- Gene
- SIX4
- Gene Name
- SIX homeobox 4
View Full Product Details