Product Details

SNP ID
rs142655621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101805977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAAGATGATTTATGACCTTTT[C/T]AGGAAGCACAAGACCATCTCTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371030 PubMed Links

Gene Details

Gene
LOC105371030
Gene Name
uncharacterized LOC105371030
There are no transcripts associated with this gene.

Gene
OR4F6
Gene Name
olfactory receptor family 4 subfamily F member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005326.1 258 Silent Mutation TTC,TTT F86F NP_001005326.1

View Full Product Details