Product Details

SNP ID
rs151109030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64096062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCGCTTCGGAGAGACTGCCTCCG[C/T]CCTTCCCCGGCCTGGAGCCGGAGTC
Phenotype
MIM: 601272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM96A PubMed Links

Gene Details

Gene
FAM96A
Gene Name
family with sequence similarity 96 member A
There are no transcripts associated with this gene.

Gene
SNX1
Gene Name
sorting nexin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242933.1 179 Missense Mutation CCC,TCC P17S NP_001229862.1
NM_003099.4 179 Missense Mutation CCC,TCC P17S NP_003090.2
NM_148955.3 179 Missense Mutation CCC,TCC P17S NP_683758.1
XM_005254611.2 179 Intron XP_005254668.1

View Full Product Details