Product Details

SNP ID
rs149744090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22110952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCTGTCCTTTGGCACCAATG[C/T]CGGGTCCCTCTGGCCAGACCCCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWA3A PubMed Links

Gene Details

Gene
VWA3A
Gene Name
von Willebrand factor A domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173615.4 967 Missense Mutation GCC,GTC A216V NP_775886.3
XM_011545742.2 967 Missense Mutation GCC,GTC A234V XP_011544044.1
XM_011545744.2 967 Intron XP_011544046.1
XM_011545745.1 967 Intron XP_011544047.1
XM_011545746.2 967 Intron XP_011544048.1
XM_017022947.1 967 Missense Mutation GCC,GTC A234V XP_016878436.1
XM_017022948.1 967 Intron XP_016878437.1
XM_017022949.1 967 Intron XP_016878438.1

View Full Product Details