Product Details

SNP ID
rs141712070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35352602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAAGAGCTCATACTTATAGTG[C/T]TCCACTTCTTTTGCGGTGCTGACAA
Phenotype
MIM: 614953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLFN11 PubMed Links

Gene Details

Gene
SLFN11
Gene Name
schlafen family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104587.1 2795 Intron NP_001098057.1
NM_001104588.1 2795 Intron NP_001098058.1
NM_001104589.1 2795 Intron NP_001098059.1
NM_001104590.1 2795 Intron NP_001098060.1
NM_152270.3 2795 Intron NP_689483.3
XM_005258068.3 2795 Silent Mutation GAA,GAG E820E XP_005258125.1
XM_006722171.3 2795 Silent Mutation GAA,GAG E820E XP_006722234.1
XM_011525465.2 2795 Silent Mutation GAA,GAG E820E XP_011523767.1
XM_017025299.1 2795 Silent Mutation GAA,GAG E820E XP_016880788.1

View Full Product Details