Product Details

SNP ID
rs142426190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44207562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCTTCTTCATCCTCGTCCTC[A/G]TCATCCTCATCCTCTTCATCATCCT
Phenotype
MIM: 600673
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN7L3 PubMed Links

Gene Details

Gene
ATXN7L3
Gene Name
ataxin 7 like 3
There are no transcripts associated with this gene.

Gene
LOC101926967
Gene Name
uncharacterized LOC101926967
There are no transcripts associated with this gene.

Gene
MIR6782
Gene Name
microRNA 6782
There are no transcripts associated with this gene.

Gene
UBTF
Gene Name
upstream binding transcription factor, RNA polymerase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076683.1 2297 Silent Mutation NP_001070151.1
NM_001076684.2 2297 Silent Mutation NP_001070152.1
NM_014233.3 2297 Silent Mutation NP_055048.1
XM_006722059.3 2297 Silent Mutation XP_006722122.1
XM_006722060.2 2297 Intron XP_006722123.1
XM_006722061.2 2297 Intron XP_006722124.1
XM_017024995.1 2297 Silent Mutation XP_016880484.1
XM_017024996.1 2297 Silent Mutation XP_016880485.1
XM_017024997.1 2297 Silent Mutation XP_016880486.1
XM_017024998.1 2297 Silent Mutation XP_016880487.1
XM_017024999.1 2297 Silent Mutation XP_016880488.1
XM_017025000.1 2297 Silent Mutation XP_016880489.1
XM_017025001.1 2297 Silent Mutation XP_016880490.1
XM_017025002.1 2297 Silent Mutation XP_016880491.1
XM_017025003.1 2297 Silent Mutation XP_016880492.1
XM_017025004.1 2297 Missense Mutation XP_016880493.1

View Full Product Details