Product Details

SNP ID
rs143918266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28612639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTAATGAAAAGGTGCCGTTTTC[C/T]CTGCCAGCTGTCCCCCAAGCTGTGT
Phenotype
MIM: 610664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0100 PubMed Links

Gene Details

Gene
KIAA0100
Gene Name
KIAA0100
There are no transcripts associated with this gene.

Gene
SGK494
Gene Name
uncharacterized serine/threonine-protein kinase SgK494
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174103.1 558 Missense Mutation AGA,GGA R176G NP_001167574.1
Gene
SPAG5-AS1
Gene Name
SPAG5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details