Product Details

SNP ID
rs149698960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6457630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTACTTCTTGGTGGGCCGGCC[C/T]ACAATGAAGATCTGGGAGGCAGGCA
Phenotype
MIM: 608921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM64A PubMed Links

Gene Details

Gene
FAM64A
Gene Name
family with sequence similarity 64 member A
There are no transcripts associated with this gene.

Gene
PITPNM3
Gene Name
PITPNM family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165966.1 2734 Silent Mutation GTA,GTG V825V NP_001159438.1
NM_031220.3 2734 Silent Mutation GTA,GTG V861V NP_112497.2
XM_011524015.2 2734 Intron XP_011522317.1
XM_011524016.2 2734 Intron XP_011522318.1
XM_011524017.2 2734 Intron XP_011522319.1

View Full Product Details