Product Details

SNP ID
rs150521243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35352769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCTGGGACCATTCGGCTTCAG[A/G]AAATACCTCGAGGCACCCAGTGGGG
Phenotype
MIM: 614953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLFN11 PubMed Links

Gene Details

Gene
SLFN11
Gene Name
schlafen family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104587.1 2628 Intron NP_001098057.1
NM_001104588.1 2628 Intron NP_001098058.1
NM_001104589.1 2628 Intron NP_001098059.1
NM_001104590.1 2628 Intron NP_001098060.1
NM_152270.3 2628 Intron NP_689483.3
XM_005258068.3 2628 Missense Mutation CCT,TCT P765S XP_005258125.1
XM_006722171.3 2628 Missense Mutation CCT,TCT P765S XP_006722234.1
XM_011525465.2 2628 Missense Mutation CCT,TCT P765S XP_011523767.1
XM_017025299.1 2628 Missense Mutation CCT,TCT P765S XP_016880788.1

View Full Product Details