Product Details
- SNP ID
-
rs150921642
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:3216271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTACTATGCATCATTGTCTCCTA[C/T]ATTCGAGTCTTCTCCACAGTCTTCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR1A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17762735] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR1A1
- Gene Name
- olfactory receptor family 1 subfamily A member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014565.2 |
651 |
Silent Mutation |
TAC,TAT |
Y217Y |
NP_055380.2 |
View Full Product Details