Product Details

SNP ID
rs138750900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54599268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACCCTCAGCCCACCACAAAAC[A/T]AGTCCGATTCCAAGGCTGGTGAGTG
Phenotype
MIM: 604810
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LILRA1 PubMed Links

Gene Details

Gene
LILRA1
Gene Name
leukocyte immunoglobulin like receptor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278318.1 1342 Intron NP_001265247.1
NM_001278319.1 1342 Nonsense Mutation AAG,TAG K432* NP_001265248.1
NM_006863.3 1342 Intron NP_006854.1

View Full Product Details