Product Details

SNP ID
rs139242998
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1000718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTGACCCGCGGCCTGTGCCAGG[A/C]GCTGGTGCCTCCGGGCGTGGCGGCC
Phenotype
MIM: 606651 MIM: 611011
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GRIN3B PubMed Links

Gene Details

Gene
GRIN3B
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138690.2 300 Missense Mutation GAG,GCG E94A NP_619635.1
XM_017026243.1 300 Intron XP_016881732.1
Gene
LOC105372235
Gene Name
uncharacterized LOC105372235
There are no transcripts associated with this gene.

Gene
TMEM259
Gene Name
transmembrane protein 259
There are no transcripts associated with this gene.

Gene
WDR18
Gene Name
WD repeat domain 18
There are no transcripts associated with this gene.

View Full Product Details