Product Details

SNP ID
rs140507040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38251392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGGGAGGGGCTTGGCTGGCGGA[A/G]GCCGGGCTGCCTGTGGAGGCCTTGA
Phenotype
MIM: 608153
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R14A PubMed Links

Gene Details

Gene
PPP1R14A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243947.1 622 Missense Mutation CTC,TTC L97F NP_001230876.1
NM_033256.2 622 Missense Mutation CTC,TTC L124F NP_150281.1

View Full Product Details