Product Details

SNP ID
rs142520824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19213188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTGGGACCCCAGAACCCAGCC[A/G]TCGCAATAGCTGATGGACATTCTGA
Phenotype
MIM: 600826 MIM: 608719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCAN PubMed Links

Gene Details

Gene
NCAN
Gene Name
neurocan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004386.2 Intron NP_004377.2
Gene
NR2C2AP
Gene Name
nuclear receptor 2C2 associated protein
There are no transcripts associated with this gene.

View Full Product Details