Product Details

SNP ID
rs144078033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8066193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGCTCCAGGATGCGCTCGGCC[C/T]GGCCCAGGTGTGAGAGGTTCAGGCC
Phenotype
MIM: 602565 MIM: 608529
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL25 PubMed Links

Gene Details

Gene
CCL25
Gene Name
C-C motif chemokine ligand 25
There are no transcripts associated with this gene.

Gene
FBN3
Gene Name
fibrillin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321431.1 8296 Missense Mutation NP_001308360.1
NM_032447.4 8296 Missense Mutation NP_115823.3
XM_017027372.1 8296 Missense Mutation XP_016882861.1
XM_017027373.1 8296 Missense Mutation XP_016882862.1
XM_017027374.1 8296 Missense Mutation XP_016882863.1
XM_017027375.1 8296 Missense Mutation XP_016882864.1
XM_017027376.1 8296 Missense Mutation XP_016882865.1
XM_017027377.1 8296 Missense Mutation XP_016882866.1
XM_017027378.1 8296 Missense Mutation XP_016882867.1
XM_017027379.1 8296 Missense Mutation XP_016882868.1

View Full Product Details