Product Details

SNP ID
rs145358114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:843597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTTGATCCACCCCAGCTTCGT[G/T]CTGACGGCCGCGCACTGCCTGCGGG
Phenotype
MIM: 130130 MIM: 177020
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELANE PubMed Links

Gene Details

Gene
ELANE
Gene Name
elastase, neutrophil expressed
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 242 Silent Mutation GTG,GTT V66V NP_002768.3
XM_011528136.1 242 Silent Mutation GTG,GTT V66V XP_011526438.1

View Full Product Details