Product Details

SNP ID
rs145729167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35550618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTGAAGGCAGTCGTCCCTCTA[A/G]TAGTAGAGTTCCTGGTCCCACCAGC
Phenotype
MIM: 137216 MIM: 609169 MIM: 613585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP4A PubMed Links

Gene Details

Gene
ATP4A
Gene Name
ATPase H+/K+ transporting alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000704.2 3134 Silent Mutation TAC,TAT Y1035Y NP_000695.2
Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
There are no transcripts associated with this gene.

Gene
TMEM147
Gene Name
transmembrane protein 147
There are no transcripts associated with this gene.

Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details